Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Multichannel Cartridge Pump Head with Reduced Pulsation for Microbore 2-stop Tubing, 12-channels, 8-rollers
Catalog No. NC1762014
Encompass
Each pump head system is composed of a heavy-duty pump head, cartridges, and tubing¿each optimal preset tubing retention (order cartridges separately). They feature stainless steel rotors, Rulon rollers, and PSF ends. Occlusion is offset, tangential, and adjustable. For small cartridges, optimum retention is preset¿simply load tubing and start pumping. Recommended for use with small cartridge 07519-85 and microbore 2-stop tubing sizes.
Catalog No. NC1762014 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0751925
Each pump head system is composed of a heavy-duty pump head, cartridges, and tubing¿each optimal preset tubing retention (order cartridges separately). They feature stainless steel rotors, Rulon rollers, and PSF ends. Occlusion is offset, tangential, and adjustable. For small cartridges, optimum retention is preset¿simply load tubing and start pumping. Recommended for use with small cartridge 07519-85 and microbore 2-stop tubing sizes.