Learn More
RWD LIFE SCIENCE INC MULTICHANNEL FIBER PHOTOMETRY
SDP
Catalog No. NC3404371
Encompass_Preferred
R821 tricolor multichannel fiber photometry system is compatible with optoelectronics equipment. It has different wavelengths of excited light, 410nm, 470nm or 560nm, of which 410nm is used to acquire reference signal and eliminate noise. The system can record signal of green fluorescence indicator like GCaMP and dLight or neurotransmitter probe and red fluorescence indicator like RCaMP, jrGECO1a or neurotransmitter probe.And also it can be applied to record the neural signal for 9 sites at the same time.
Catalog No. NC3404371 Supplier RWD LIFE SCIENCE INC Supplier No. R821
Add to Cart
Add to Cart
Request a Quote
R821 tricolor multichannel fiber photometry system is compatible with optoelectronics equipment. It has different wavelengths of excited light, 410nm, 470nm or 560nm, of which 410nm is used to acquire reference signal and eliminate noise. The system can record signal of green fluorescence indicator like GCaMP and dLight or neurotransmitter probe and red fluorescence indicator like RCaMP, jrGECO1a or neurotransmitter probe.And also it can be applied to record the neural signal for 9 sites at the same time.