Learn More
World Precision Instrument MultiChannel Perfusion System
SDP
Catalog No. 502538672
Encompass
  • Fast solenoid valve (response time 2ms)
  • Programmable PC control with user-friendly GUI interface (software included)
  • Manual
  • 8-Channels-10ml syringes
  • Gravity feed
  • Quick cleaning and priming valves included
Catalog No. 50-253-8672 Supplier World Precision Instrument Supplier No. MPS4
  • Fast solenoid valve (response time 2ms)
  • Programmable PC control with user-friendly GUI interface (software included)
  • Manual
  • 8-Channels-10ml syringes
  • Gravity feed
  • Quick cleaning and priming valves included