Learn More
SPEX CERTIPREP LLC 3P MULTICOMPONENT ANION STD 100ML
Catalog No. NC9831417
Encompass_Preferred
(HPS by Zeptometrix) Multicomponent Anion Standard in H2O; 7 components at 100 ug/mL packaged in 2 solutions; 12 month expiry date. Sol A- Br, Cl, F, NO3, PO4, SO4. Sol B-NO2
Catalog No. NC9831417 Supplier SPEX CERTIPREP LLC 3P Supplier No. IC1100
(HPS by Zeptometrix) Multicomponent Anion Standard in H2O; 7 components at 100 ug/mL packaged in 2 solutions; 12 month expiry date. Sol A- Br, Cl, F, NO3, PO4, SO4. Sol B-NO2