Learn More
Analytical Sales & Services MULTICON Gen II Heater Sleeve Controller, Touchscreen Interface, Up to 250 W Heater Power, Unit
Catalog No. NC2876265
Encompass
The MULTICON Gen II Heater Sleeve Controller offers precise temperature management for HPLC columns, enhancing operational efficiency with its user-friendly touchscreen and real-time data display. Designed for high-power operation, it supports temperatures from 5ºC above ambient to 130ºC, powering heaters up to 250 W. Its stacking enclosure conserves lab space while USB and Modbus RS485 connectivity enable integration of up to 31 controllers, making it a versatile solution for laboratory environments.
Catalog No. NC2876265 Supplier Analytical Sales & Services Supplier No. MULTICON
Add to Cart
Add to Cart
Request a Quote
The MULTICON Gen II Heater Sleeve Controller offers precise temperature management for HPLC columns, enhancing operational efficiency with its user-friendly touchscreen and real-time data display. Designed for high-power operation, it supports temperatures from 5ºC above ambient to 130ºC, powering heaters up to 250 W. Its stacking enclosure conserves lab space while USB and Modbus RS485 connectivity enable integration of up to 31 controllers, making it a versatile solution for laboratory environments.