Learn More
Alkali Scientific MultiCycler 332 MultiBlock The
SDP
Catalog No. 503029662
Encompass_Preferred
Double or triple block format Intuitive touch screen control Program Wizard Gradient capabilities Alkali ScientificT3-0332G is designed for laboratories wanting flexibility in their applications Alkalis MultiCyclerThermal Cyclers offer multiple independent cycling blocks in a single unit Dual or triple block models are available both offering extremely fast cycling profiles large touch screen interfaces self-adjusting heated lids gradient capabilities and an easy-to-use program wizard All programming features are accessed from the large 10 full color touch screen The screen can be angled towards the user when in use and then tucked away to save space A program wizard guides users through the simple protocol set up process Advanced programming features include gradient adjustable ramp rates and time increments and decrements For multi-user labs password protection prevents interference of an existing run A USB provides for unlimited program storage and easy transfer of protocols
Catalog No. 50-302-9662 Supplier Alkali Scientific Supplier No. RD0332G
Add to Cart
Add to Cart
Request a Quote
Double or triple block format Intuitive touch screen control Program Wizard Gradient capabilities Alkali ScientificT3-0332G is designed for laboratories wanting flexibility in their applications Alkalis MultiCyclerThermal Cyclers offer multiple independent cycling blocks in a single unit Dual or triple block models are available both offering extremely fast cycling profiles large touch screen interfaces self-adjusting heated lids gradient capabilities and an easy-to-use program wizard All programming features are accessed from the large 10 full color touch screen The screen can be angled towards the user when in use and then tucked away to save space A program wizard guides users through the simple protocol set up process Advanced programming features include gradient adjustable ramp rates and time increments and decrements For multi-user labs password protection prevents interference of an existing run A USB provides for unlimited program storage and easy transfer of protocols