Learn More
IW Tremont Co Inc LabExact Nonsterile MultiDensity Syringe Filters,1.5um, 25mm, 100 pack
SDP
Catalog No. 502237843
Encompass_Preferred
LabExact Disposable non-sterile MultiDensity Nano Glass Microfiber syringe filters have several layers of density that speeds and optimizes filtration. The proprietary media includes multi pre-filtration stages and final retention stage down to 1.5um. Built-in prefilter captures particulate from >300?m and binds typically difficult to filter substances without clogging such as lipids (natural oils and waxes), petrochemical oils, tars, cellular matter and proteins.Filters are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. The filters feature Luer-Lok and pressure fitting Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The unique concentric funnel shaped barrel increases volume and decreases back pressure.
This item is not returnable. View return policy
Catalog No. 50-223-7843 Supplier IW Tremont Co Inc Supplier No. LEIWTES10705
LabExact Disposable non-sterile MultiDensity Nano Glass Microfiber syringe filters have several layers of density that speeds and optimizes filtration. The proprietary media includes multi pre-filtration stages and final retention stage down to 1.5um. Built-in prefilter captures particulate from >300?m and binds typically difficult to filter substances without clogging such as lipids (natural oils and waxes), petrochemical oils, tars, cellular matter and proteins.Filters are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. The filters feature Luer-Lok and pressure fitting Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The unique concentric funnel shaped barrel increases volume and decreases back pressure.