Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences MultiDish 6 Well Round, 35 mm wells, Sterile, 2.0-5.0 mL Working volume
SDP

Catalog No. 50930286
Encompass

MultiDish Surface is useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™Δ certified.
Well size: 35 mm
Working volume: 2.0-5.0 mL
Area: 9.6 cm2
Plate size: 88 x 132 mm

Catalog No. 50-930-286 Supplier Electron Microscopy Sciences Supplier No. 70428
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MultiDish Surface is useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™Δ certified.
Well size: 35 mm
Working volume: 2.0-5.0 mL
Area: 9.6 cm2
Plate size: 88 x 132 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.