Learn More
Cayman Chemical MultIdrUG ResIstnce asy KI 1ea
SDP
Catalog No. 502895234
Encompass_Preferred
Cayman's Multidrug Resistance Assay Kit provides a convenient tool for studying the modulation of cellular multidrug resistance (MDR) machinery. The kit employs calcein AM, a cell-permeable nonfluorescent dye, which is cleaved intracellularly to yield the impermeant fluorescent molecule, calcein. This calcein is rapidly excluded from cells expressing the MDR proteins P-gp or MRP, and so functions as a probe for the detection of chemical compounds inhibiting MDR proteins. Propidium iodide is provided as a stain for marking dead or damaged cells. Cyclosporin A, a competitive inhibitor of MDR proteins, and verapamil, a noncompetitive inhibitor, are included as controls.
Catalog No. 50-289-5234 Supplier Cayman Chemical Supplier No. 6003701 EA
Cayman's Multidrug Resistance Assay Kit provides a convenient tool for studying the modulation of cellular multidrug resistance (MDR) machinery. The kit employs calcein AM, a cell-permeable nonfluorescent dye, which is cleaved intracellularly to yield the impermeant fluorescent molecule, calcein. This calcein is rapidly excluded from cells expressing the MDR proteins P-gp or MRP, and so functions as a probe for the detection of chemical compounds inhibiting MDR proteins. Propidium iodide is provided as a stain for marking dead or damaged cells. Cyclosporin A, a competitive inhibitor of MDR proteins, and verapamil, a noncompetitive inhibitor, are included as controls.