Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multielement Ion Chromatography Anion Standard Solution, Certified for ion chromatography, MilliporeSigma™ Supelco™

Catalog No. 111004066 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4066 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4066 Supplier MilliporeSigma Supelco Supplier No. 8988650MLF
Add to Cart
Add to Cart

Traceable to SI, 10.0 mg/kg (0.2 F-, Cl-, Br--, NO3-, PO43-, SO42- each anion)

Certified multiion standard for Ion Chromatography: high purity starting materials
TRUSTED_SUSTAINABILITY

Specifications

Synonym Certified Multielement Ion Chromatography Anion Standard Solution
Quantity 50 mL
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.