Learn More
Perkin Elmer US LLC Multi-Element Standard B -MES110901A, 10% HNO3, 125 mL
Catalog No. 502117327
Encompass_Preferred
Details Concentration Element 10,000 µg/mL Ca, Mg, P, K, Na
Catalog No. 50-211-7327 Supplier Perkin Elmer US LLC Supplier No. N9308546
Details Concentration Element 10,000 µg/mL Ca, Mg, P, K, Na