Learn More
DWK Life Sciences Kimble™ Multi-Fiber Test Fabric, 36x4
SDP
Catalog No. k7480300049 Shop All DWK Life Sciences Products
Click to view available options
:
Multi-Fiber Test Fabric, 36x4
Catalog No. K7480300049
Supplier: DWK Life Sciences 7480300049
Only null left
Add to Cart
Add to Cart
This test fabric is useful for testing dyes on a variety of fabrics.
  • Accessory for Microscale Williamson Kits
  • Three-foot length of fabric four inches wide
  • 8 strips of different fibers woven side by side: Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-fiber Test Fabric
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.