Learn More
Siemens Healthineers Multifibren™ U Reagent
Catalog No. 23044709 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 2 mL
10 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-709 10 x 2 mL
23-044-710 10 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-709 Supplier Siemens Healthineers Supplier No. 10446689
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of fibrinogen in plasma.
Multifibren™ U Reagent is a bovine thrombin reagent used in the modified Clauss determination of fibrinogen for the detection of hereditary or acquired hypo- and hyperfibrinogenemia and dysfibrinogenemia. The reagent is insensitive to heparin up to 2.0 U/mL and has a wide measuring range.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Fibrinogen Testing
Quantity 10 x 2 mL