Learn More
Research Products International Corp MultiFlex Pipet Tips for Gel Loading, Non-Sterile, RNase and DNase Free, 0.5-200ul, Flat Tip, 200 per Package
SDP
Catalog No. 50550204
Encompass
MultiFlex tips are designed for precise loading of gels run in horizontal and vertical electrophoresis equipment. Other applications include DNA sequencing, nick translations, restriction digests and radioactive labelling. Revolutionary design provides a tight positive seal on small volume (1-200µl) sizes of air displacement micro-pipettors. MultiFlex tips fit all popular micro-pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA and Volac.
Tips are constructed of nucleated polypropylene and can be autoclaved to 120°C.
Catalog No. 50-550-204 Supplier Research Products International Corp Supplier No. 148600
MultiFlex tips are designed for precise loading of gels run in horizontal and vertical electrophoresis equipment. Other applications include DNA sequencing, nick translations, restriction digests and radioactive labelling. Revolutionary design provides a tight positive seal on small volume (1-200µl) sizes of air displacement micro-pipettors. MultiFlex tips fit all popular micro-pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA and Volac.
Tips are constructed of nucleated polypropylene and can be autoclaved to 120°C.