Learn More
Andwin Scientific MULTIFLEX PIPET TIPS
SDP
Catalog No. NC9770832
Encompass_Preferred
MultiFlex Pipet Tips, Rounded end, 0.57mm O.D. Length: 83mm 5microliter graduation. Positive displacement accuracy using air displacement pipettors. 33mm flexible microcapillary allows precision pipetting in the 0.5 to 200microliter range.
Catalog No. NC9770832 Supplier Andwin Scientific Supplier No. 53550167
MultiFlex Pipet Tips, Rounded end, 0.57mm O.D. Length: 83mm 5microliter graduation. Positive displacement accuracy using air displacement pipettors. 33mm flexible microcapillary allows precision pipetting in the 0.5 to 200microliter range.