Learn More
Cayman Chemical Multiflorenol, Purity: ≥85%, CAS: 2270-62-4, 5 mg
SDP
Catalog No. 502988196
Encompass_Preferred
A triterpene; inhibits TPA-induced EBV-EA activation in vitro in a concentration-dependent manner
Catalog No. 50-298-8196 Supplier Cayman Chemical Supplier No. 297095 MG
A triterpene; inhibits TPA-induced EBV-EA activation in vitro in a concentration-dependent manner