Learn More
Litron Laboratories MultiFlow DNA Damage Kit, p53, γH2AX, Phospho-Histone H3, 96-well plate format, up to 384 samples
Catalog No. NC1306275
Encompass
MultiFlow DNA Damage Kit, p53, γH2AX, Phospho-Histone H3, 96-well plate format, up to 384 samples
Catalog No. NC1306275 Supplier Litron Laboratories Supplier No. MULTIFLOW(P53,YH2AX,H3)
MultiFlow DNA Damage Kit, p53, γH2AX, Phospho-Histone H3, 96-well plate format, up to 384 samples