Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Multifunctional Tool for M210 and BMP™21 Label Printers

Catalog No. 19000750 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-750 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-000-750 Supplier Brady™ Supplier No. M210TOOL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this multifunctional tool to operate your label printer with one hand, leaving your other hand free.

The multifunctional accessory for Brady™ Label Printers consists of a magnet, flashlight, stand, belt loop and pole strap.

  • Ideal for an on-the-go labeling environment
  • Built in LED flashlight helps you create labels in low-lit areas
  • 2 AAA batteries required (not included)
  • Compatible with Brady™ BMP™21, BMP™21-LAB, M210, and M210-LAB Label Printers

Specifications

Color Black
For Use With (Equipment) BMP21 Label Printer, Brady M210 Label Printer
Includes Magnet, Flashlight, Stand, Belt Loop, Pole Strap
Certifications/Compliance 2011/65/EU and 2015/863
Alternate Part Number M210-TOOL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.