Learn More
Electron Microscopy Sciences Ilford Multigrade IV RC Paper, 5x7
SDP
Supplier:  Electron Microscopy Sciences 8302025
Encompass
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m 2)with a resin coated base.
Catalog No. 50-313-59