Learn More
Electron Microscopy Sciences Ilford Multigrade IV RC Paper, 8x10
SDP
Catalog No. 5031360
Encompass
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m 2)with a resin coated base.
Catalog No. 50-313-60 Supplier Electron Microscopy Sciences Supplier No. 830211M
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m 2)with a resin coated base.