Learn More
Electron Microscopy Sciences Ilford Multigrade RC Cooltone, 8 x 10
SDP
Catalog No. 5031367
Encompass
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers. Premium Quality, Cool Tone – Variable contrast. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is equally suitable for printing all black and white negatives including chromogenic negatives, such as ILFORD XP2 SUPER.
Catalog No. 50-313-67 Supplier Electron Microscopy Sciences Supplier No. 830301C
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers. Premium Quality, Cool Tone – Variable contrast. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is equally suitable for printing all black and white negatives including chromogenic negatives, such as ILFORD XP2 SUPER.