Learn More
Electron Microscopy Sciences Ilford Multigrade RC Express Pf Paper, 10" 250 FT
SDP
Catalog No. 5031362
Encompass
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast for high speed printing applications. MULTIGRADE RC Express PF has the standard weight of (190g/m 2) with a resin coated base. It is especially designed for high speed printing applications, where automatic and semiautomatic printers, printer-processors and minilabs are used.
Size: 10"
Catalog No. 50-313-62 Supplier Electron Microscopy Sciences Supplier No. 8302410
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast for high speed printing applications. MULTIGRADE RC Express PF has the standard weight of (190g/m 2) with a resin coated base. It is especially designed for high speed printing applications, where automatic and semiautomatic printers, printer-processors and minilabs are used.
Size: 10"