Learn More
Electron Microscopy Sciences Ilford Multigrade RC Warmtone Paper, 8x10
SDP
Supplier:  Electron Microscopy Sciences 830271C
Encompass
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers. Premium Quality, Warm Tone – Variable Contrast. MULTIGRADE RC Warmtone Paper has the standard weight of(190g/m 2) with a resin coated base. It is equally suitable for printing conventional black and white and XP2 SUPER negatives.
Catalog No. 50-313-64