Learn More
Andwin Scientific MULTIGUARD TIP,ST,10UL, 5PK/CS
SDP
Catalog No. NC0430765
Encompass_Preferred
The 10 μL Sorenson™ low binding aerosol barrier tips are recommended for molecular biology applications, such as PCR. This product has a new surface technology designed for quantitative assays in which sample binding can significantly change results or for critical small volume protocols that require a high degree of accuracy. The tips are sterile and are sold in a case of 5 packs.
Catalog No. NC0430765 Supplier Andwin Scientific Supplier No. 38000T
The 10 μL Sorenson™ low binding aerosol barrier tips are recommended for molecular biology applications, such as PCR. This product has a new surface technology designed for quantitative assays in which sample binding can significantly change results or for critical small volume protocols that require a high degree of accuracy. The tips are sterile and are sold in a case of 5 packs.