Learn More
Electron Microscopy Sciences Multi Holder for 8 Pin Stubs, Zeiss with Built-in Dov
SDP
Catalog No. 501931035
Encompass
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: Zeiss with Built-in Dovetail
Catalog No. 50-193-1035 Supplier Electron Microscopy Sciences Supplier No. 7595208ZE
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: Zeiss with Built-in Dovetail