Learn More
Electron Microscopy Sciences Multi Holder for 8 Pins, Hitachi M4
SDP
Catalog No. 501901515
Encompass
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: Hitachi M4
Catalog No. 50-190-1515 Supplier Electron Microscopy Sciences Supplier No. 7595208H
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: Hitachi M4