Learn More
Electron Microscopy Sciences Multimagstir Genie, 120 V/100 V
SDP
Supplier:  Electron Microscopy Sciences 7240512
Encompass_Preferred
The revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles. It can be used with up to 4 1L flasks/beakers or 3 1L spinner flasks and a maximum sample weight of 10.0 kg. Three power level, a speed range of 10-1200- RPM and reverse and interval stirring make the stirrer ideal for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers.
Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction.
Manual mode provides stir, stir reverse, and time functions.
Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to 9 distinct steps. Up to 4 stirring programs can be saved.
Automatic mode recalls and executes one of up to 4 user-defined protocols.
Catalog No. 50-189-9886