Learn More
Andwin Scientific MULTIMAGSTIR GENIE 120V/100V
SDP
Catalog No. 50949854
Encompass_Preferred
The revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles. Microprocessor controls offer precision and reproducibility without internal heat build-up. DxWxH 14.8 x 12.8 x 1.9in.
Catalog No. 50-949-854 Supplier Andwin Scientific Supplier No. 7240512
The revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles. Microprocessor controls offer precision and reproducibility without internal heat build-up. DxWxH 14.8 x 12.8 x 1.9in.