Learn More
Cole-Parmer Fluke 116 HVAC Digital Multimeter w/ Temperature
Catalog No. 50992387 Shop All Cole-Parmer Products
Encompass
This auto-ranging multimeter measures temperature with included type K thermocouple probe, tests flame rod circuits with microamp function, records min/max measurements over 100-hour period with time stamp, and offers voltage check for stray voltages. Meter also features auto-off and a low-battery indicator. The Fluke 116 was specifically designed for the HVAC professional. It has everything needed in an HVAC meter including temperature and microamp measurements to quickly troubleshoot problems with HVAC equipment and flame sensors. Use the Fluke 116 to test motor start and run capacitors up to 10,000 microfarads. Rely on the Fluke 116 to make your everyday measurements - up to 600 volts AC or DC, 40 MÙ, diode test and exceptionally fast continuity.
Catalog No. 50-992-387 Supplier Cole-Parmer Supplier No. 2601620
This auto-ranging multimeter measures temperature with included type K thermocouple probe, tests flame rod circuits with microamp function, records min/max measurements over 100-hour period with time stamp, and offers voltage check for stray voltages. Meter also features auto-off and a low-battery indicator. The Fluke 116 was specifically designed for the HVAC professional. It has everything needed in an HVAC meter including temperature and microamp measurements to quickly troubleshoot problems with HVAC equipment and flame sensors. Use the Fluke 116 to test motor start and run capacitors up to 10,000 microfarads. Rely on the Fluke 116 to make your everyday measurements - up to 600 volts AC or DC, 40 MÙ, diode test and exceptionally fast continuity.