Learn More
Cole-Parmer Extech 380941 DC/AC Clamp Meter, 200 Amp
Catalog No. 50993003 Shop All Cole-Parmer Products
Encompass
This clamp meter amp features a large 4,000 count LCD display with full function indication, along with a fast 40 segment analog bargraph. Its One touch "Auto Zero" provides DCA measurements to eliminate the need to turn a knob to zero adjust. There also is a data hold, maximum and minimum recall display. The opening of the clamp jaw is about 0.9 inches or 23 millimeters.
Catalog No. 50-993-003 Supplier Cole-Parmer Supplier No. 2684882
This clamp meter amp features a large 4,000 count LCD display with full function indication, along with a fast 40 segment analog bargraph. Its One touch "Auto Zero" provides DCA measurements to eliminate the need to turn a knob to zero adjust. There also is a data hold, maximum and minimum recall display. The opening of the clamp jaw is about 0.9 inches or 23 millimeters.