Learn More
Cole-Parmer Extech 380942 True RMS AC/DC Clamp Meter, 30A
Supplier:  Cole-Parmer 2684884
Encompass
This clamp meter amp features a large 4,000 count LCD display with full function indication, along with a fast 40 segment analog bargraph. Its One touch "Auto Zero" provides DCA measurements to eliminate the need to turn a knob to zero adjust. There also is a data hold, maximum and minimum recall display. The opening of the clamp jaw is about 0.9 inches or 23 millimeters.
Catalog No. 50-993-004