Learn More
Grainger REED Instruments True RMS Industrial Multimeter, Bluetooth, Data Logger, Waterproof
Catalog No. 502763497
Encompass_Preferred
The industrial-grade True RMS multimeter is a robust, data-logging instrument designed for accurate electrical measurements in demanding environments. Featuring a waterproof IP67-rated double molded housing, this device is built for durability at challenging work sites. Users can analyze signals via a high-resolution 50,000 count, 320 x 240 TFT color display and monitor trends.
  • True RMS measurement for accurate non-sinusoidal signal analysis
  • Wireless Bluetooth connectivity for remote data logging and trend review
  • Waterproof IP67-rated double molded housing for harsh environments
  • Safety rated for CAT IV 600 V and CAT III 1,000 V
  • Large 50,000 count, 320 x 240 TFT color backlit display
  • Includes test leads, type K thermocouple wire probe, thermocouple adapter, weatherproof plugs, AC adapter, Li-ion rechargeable battery, and software
Catalog No. 50-276-3497 Supplier Grainger Supplier No. 161C82
The industrial-grade True RMS multimeter is a robust, data-logging instrument designed for accurate electrical measurements in demanding environments. Featuring a waterproof IP67-rated double molded housing, this device is built for durability at challenging work sites. Users can analyze signals via a high-resolution 50,000 count, 320 x 240 TFT color display and monitor trends.
  • True RMS measurement for accurate non-sinusoidal signal analysis
  • Wireless Bluetooth connectivity for remote data logging and trend review
  • Waterproof IP67-rated double molded housing for harsh environments
  • Safety rated for CAT IV 600 V and CAT III 1,000 V
  • Large 50,000 count, 320 x 240 TFT color backlit display
  • Includes test leads, type K thermocouple wire probe, thermocouple adapter, weatherproof plugs, AC adapter, Li-ion rechargeable battery, and software