Learn More
Cayman Chemical Multi-Parameter Apoptosis Assay Kit, 1 ea
SDP
Catalog No. 502894276
Encompass_Preferred
Cayman's Multi-Parameter Apoptosis Assay Kit is a simple way to probe for multiple different apoptotic readouts at the same time. This kit employs FITC-conjugated Annexin V as a probe for phosphatidylserine on the outer membrane of apoptotic cells, TMRE as a probe for Δψm, RedDotTM2 as an indicator of plasma membrane permeability/cell viability, and Hoechst Dye to demonstrate nuclear morphology. The kit allows phenotypic characterization of multiple different cell death parameters at the single-cell level. The reagents provided in the kit are sufficient to run 100 samples.
Catalog No. 50-289-4276 Supplier Cayman Chemical Supplier No. 6012801 EA
Cayman's Multi-Parameter Apoptosis Assay Kit is a simple way to probe for multiple different apoptotic readouts at the same time. This kit employs FITC-conjugated Annexin V as a probe for phosphatidylserine on the outer membrane of apoptotic cells, TMRE as a probe for Δψm, RedDotTM2 as an indicator of plasma membrane permeability/cell viability, and Hoechst Dye to demonstrate nuclear morphology. The kit allows phenotypic characterization of multiple different cell death parameters at the single-cell level. The reagents provided in the kit are sufficient to run 100 samples.