Learn More
Echelon Biosciences Research Labs MULTIPIP GRIP 2.5 ug
SDP
Supplier:  Echelon Biosciences Research Labs G99012.5UG
Encompass
Each vial of MultiPIP Grip contains lyophilized, recombinant GST-tagged LL5-PH domain protein which has affinity for all phosphoinositide polyphosphates. This MultiPIP binding protein can be used as a control reagent with PIP Strips, PIP Arrays, and PIP Plates. It is easily reconstituted in water or buffered solutions. Each 2.5 g vial contains enough protein for use on one PIP Strip or PIP Array. MultiPIP Grip can be detected with commercial anti-GST antibodies. This item is non returnable
Catalog No. 50-720-351