Learn More
Research Products International Corp Multi-Pipettor Station
SDP
Catalog No. 50550434
Encompass
Ideal for holding most popular brands of pipettors. Pipettors are held in place by resting against upper or lower rack. Constructed of attractive acrylic for clarity and durability. The upper rack has 25mm wide slots and the lower rack has 20mm wide slots. Accommodates up to six pipettors.
Catalog No. 50-550-434 Supplier Research Products International Corp Supplier No. PH6000
Ideal for holding most popular brands of pipettors. Pipettors are held in place by resting against upper or lower rack. Constructed of attractive acrylic for clarity and durability. The upper rack has 25mm wide slots and the lower rack has 20mm wide slots. Accommodates up to six pipettors.