Learn More
Bio Rad Laboratories MULTIPLATE 48-WELL UNSKIRTED P
Catalog No. 509011610
Encompass
PCR plates, 48-well; Bio-Rad; Multiplate; 8 x 6 format; Unskirted; Natural; Thin-wall; Polypropylene; Can be cut for use with fewer samples; Accommodate PCR volumes of 5 to 125uL; Free of DNase, RNase, and human DNA; 50/Pk
Catalog No. 50-901-1610 Supplier Bio Rad Laboratories Supplier No. MLP4801
PCR plates, 48-well; Bio-Rad; Multiplate; 8 x 6 format; Unskirted; Natural; Thin-wall; Polypropylene; Can be cut for use with fewer samples; Accommodate PCR volumes of 5 to 125uL; Free of DNase, RNase, and human DNA; 50/Pk