Learn More
Bio Rad Laboratories MULTIPLATE 96-WELL UNSKIRTED P
Supplier:  Bio Rad Laboratories MLP9651
Encompass
PCR plate, 96-well; Bio-Rad; Multiplate; Unskirted; Polypropylene; White; Can be cut for use with fewer samples; Accommodate PCR volumes of 5 to 125uL; Very low protein binding; Process-sampled; Free of DNase, RNase, and human DNA; 25/Pk
Catalog No. 50-901-1902