Learn More
Bio Rad Laboratories MULTIPLATE 96-WELL UNSKIRTED P
Supplier:  Bio Rad Laboratories MLP9601
Encompass
PCR Plates, 96-Well Unskirted; Bio-Rad; Multiplate; Polypropylene; Accommodate PCR volumes of 5-125uL; Negative for DNase, RNase, human DNA; Natural; 25 plates
Catalog No. 50-901-0030