Learn More
Bio Rad Laboratories MULTIPLATE LOW-PROFILE 48-WELL
Catalog No. 509011809
Encompass
PCR plate, 48-well; Bio-Rad; Multiplate; Unskirted; Low-profile; Polypropylene; 8 x 6 format; Natural; Rigid top surface; Can be cut for use with fewer samples; For 5 to 125uL; Process-sampled; Free of DNase, RNase, and human DNA; 50/Pk
Catalog No. 50-901-1809 Supplier Bio Rad Laboratories Supplier No. MLL4801
PCR plate, 48-well; Bio-Rad; Multiplate; Unskirted; Low-profile; Polypropylene; 8 x 6 format; Natural; Rigid top surface; Can be cut for use with fewer samples; For 5 to 125uL; Process-sampled; Free of DNase, RNase, and human DNA; 50/Pk