Learn More
Grainger Multiple Appliance Interlock Control
Catalog No. 502752934
Encompass_Preferred
The Grainger website features mechanically actuated safety interlock switches. These switches are designed to help protect personnel by shutting down machinery when guards are opened. They feature direct opening mechanisms and positive-break contacts for reliable operation. Various configurations are available including non-contact, tongue-and-groove, and trapped key. These switches can be used in a wide range of applications and industries. They meet various safety standards and regulations. Multiple contact options and terminations are offered.
Catalog No. 50-275-2934 Supplier Grainger Supplier No. 14U180
The Grainger website features mechanically actuated safety interlock switches. These switches are designed to help protect personnel by shutting down machinery when guards are opened. They feature direct opening mechanisms and positive-break contacts for reliable operation. Various configurations are available including non-contact, tongue-and-groove, and trapped key. These switches can be used in a wide range of applications and industries. They meet various safety standards and regulations. Multiple contact options and terminations are offered.