Learn More
Braintree Scientific Multiple Capsule Filler,filler, tamper, spreader for size 9
SDP
Catalog No. 501956142
Encompass
MultipleCapsule Filler Fill 40 capsules at a time to withinin +/- 7-10% of average weight Includes filler, tamper, and spreader **Sold for research purposes only and not for human use**
Catalog No. 50-195-6142 Supplier Braintree Scientific Supplier No. XFILL9
MultipleCapsule Filler Fill 40 capsules at a time to withinin +/- 7-10% of average weight Includes filler, tamper, and spreader **Sold for research purposes only and not for human use**