Learn More
Bellco Glass, Inc. Multiple Cavity Reagent
SDP
Catalog No. 502329871
Encompass_Preferred
DescriptionReagent Reservoirs are designed to assist in repetitive or robotic liquid transferTrough Bottom design promotes funneling of liquid to maximize reagent recovery by pipet tipsLow profile is ideal for small volume robotic tipsMultiple wells allow for dispensing of various reagentsPolypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applicationsAutoclavable Pack Description5/BagCase Quantity25SterileNoNon-pyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume22mL/WellRNase/DNase FreeYes
Catalog No. 50-232-9871 Supplier Bellco Glass, Inc. Supplier No. 229561000
DescriptionReagent Reservoirs are designed to assist in repetitive or robotic liquid transferTrough Bottom design promotes funneling of liquid to maximize reagent recovery by pipet tipsLow profile is ideal for small volume robotic tipsMultiple wells allow for dispensing of various reagentsPolypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applicationsAutoclavable Pack Description5/BagCase Quantity25SterileNoNon-pyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume22mL/WellRNase/DNase FreeYes