Learn More
Electron Microscopy Sciences Multiple FIB Grid Holder
SDP
Catalog No. 501927130
Encompass_Preferred
Larger size enables this FIB grid holder to handle multiple FIB grids of the same thickness. Can also be used to safely store FIB grids with thin section (lamellae) attached. Convenient brass thumbscrews make loading and unloading easy. Pin: Standard 3.2 mm ( 1/8"). Material: Vacuum grade aluminum with brass thumbscrews.
Catalog No. 50-192-7130 Supplier Electron Microscopy Sciences Supplier No. 7595005
Larger size enables this FIB grid holder to handle multiple FIB grids of the same thickness. Can also be used to safely store FIB grids with thin section (lamellae) attached. Convenient brass thumbscrews make loading and unloading easy. Pin: Standard 3.2 mm ( 1/8"). Material: Vacuum grade aluminum with brass thumbscrews.