Learn More
Analytical Sales & Services MµLtisleeve 4 Port Temperature Control Unit
Catalog No. NC0119898
Encompass
*Self-calibrating * Plug & Play smart heater technology * Controls up to 4 column heaters simµLtaneously * 4 independent channels * Used in mµLtiplexing applications * Temperature range from 5C above ambient to 85c * Controller comes with cables, manual, and usb gui software.
Catalog No. NC0119898 Supplier Analytical Sales & Services Supplier No. MULTISLEEVE
*Self-calibrating * Plug & Play smart heater technology * Controls up to 4 column heaters simµLtaneously * 4 independent channels * Used in mµLtiplexing applications * Temperature range from 5C above ambient to 85c * Controller comes with cables, manual, and usb gui software.