Learn More
Discovery Life Sciences Multiple Myeloma Actv Tx PBMC
SDP
Catalog No. 5000804008
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04008 Supplier Discovery Life Sciences Supplier No. 121465
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays