Learn More
Discovery Life Sciences Multiple Myeloma BMMC
SDP
Catalog No. 5000804116
Encompass
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development
Catalog No. 50-008-04116 Supplier Discovery Life Sciences Supplier No. 100111.1
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development