Learn More
US Biomax Inc Multiple organ normal tissue array, 11 cases/24 cores, replacing BN242c
SDP
Catalog No. NC3734753 Shop All US Biomax Inc Products
Encompass
Multiple organ normal tissue microarray, containing 12 type of multiple normal organs (stomach, esophagus, colon, rectum, liver, small intestine, thyroid, spleen, tonsil, testis, bladder and larynx), duplicate cores per block
Catalog No. NC3734753 Supplier US Biomax Inc Supplier No. BN242D
Multiple organ normal tissue microarray, containing 12 type of multiple normal organs (stomach, esophagus, colon, rectum, liver, small intestine, thyroid, spleen, tonsil, testis, bladder and larynx), duplicate cores per block