Learn More
US Biomax Inc Multiple organ normal tissue array, 12 cases/24 cores, replacing BN243c
SDP
Catalog No. NC3734751 Shop All US Biomax Inc Products
Encompass
Multiple organ normal tissue microarray, containing 12 types of normal organs (cerebrum, cardiac muscle, lung, liver, uterine cervix, colon, kidney, breast, ovary, pancreas, prostate and skin), each organ taken from single normal human individuals, duplicate cores per case
Catalog No. NC3734751 Supplier US Biomax Inc Supplier No. BN243D
Multiple organ normal tissue microarray, containing 12 types of normal organs (cerebrum, cardiac muscle, lung, liver, uterine cervix, colon, kidney, breast, ovary, pancreas, prostate and skin), each organ taken from single normal human individuals, duplicate cores per case