Learn More
Andwin Scientific MULTIPLE SAMPLE HEAD SET. INCL
SDP
Catalog No. 50820880
Encompass_Preferred
Multiple sample head set. Includes 6 in. platform head, insert for 60 micro-tubes (5.0 mm to 10.5 mm) and 96-well plate insert. For use with Vortex-Genie; 2 and Vortex-Genie; 2T only.
Catalog No. 50-820-880 Supplier Andwin Scientific Supplier No. 74043010
Multiple sample head set. Includes 6 in. platform head, insert for 60 micro-tubes (5.0 mm to 10.5 mm) and 96-well plate insert. For use with Vortex-Genie; 2 and Vortex-Genie; 2T only.