Learn More
US Biomax Inc Multiple sarcoma of soft tissue array with normal tissue, including pathology grade, TNM and clinical stage, 64 cases/192 cores, replacing SO208
SDP
Catalog No. NC3433571 Shop All US Biomax Inc Products
Encompass
Multiple sarcoma of soft tissue microarray, containing 35 cases of liposarcoma, 9 dermatofibrosarcoma carina, 2 angiosarcoma, 3 leiomyosarcoma, 5 malignant schwannoma, 1 synovial sarcoma, 4 striated muscle tissue, 1 skeletal muscle tissue, 2 each of smooth muscle tissue and 2 endometrium tissue, triple cores per case
Catalog No. NC3433571 Supplier US Biomax Inc Supplier No. SO208A
Multiple sarcoma of soft tissue microarray, containing 35 cases of liposarcoma, 9 dermatofibrosarcoma carina, 2 angiosarcoma, 3 leiomyosarcoma, 5 malignant schwannoma, 1 synovial sarcoma, 4 striated muscle tissue, 1 skeletal muscle tissue, 2 each of smooth muscle tissue and 2 endometrium tissue, triple cores per case